missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Alkalinity M (alkaphot M) Test Kit

Product Code. 13171451
Click to view available options
Quantity:
1 Each
Unit Size:
250 tests
This item is not returnable. View return policy

Product Code. 13171451

Brand: Palintest™ AP 250

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Concentration 0 to 500 mg/L
Quantity 1 Each
compliance-icons
Product Identifier
  • Test kit Palintest alkalinity M (alkaphot M) re-fill packs for 250 tests 0 to 500 (CaC03)mg M
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : adipic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.