missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Alkaline Reagent

Product Code. 10729391
Change view
Click to view available options
Type:
Alkaline Reagent
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10729391 Alkaline Reagent 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10729391 Supplier WTW™ Supplier No. 150130
  • ONLINE EXCLUSIVE PRICE
    £132.00
    £128.68 / 250mL
    Save £3.32
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.

TRUSTED_SUSTAINABILITY

Specifications

Type Alkaline Reagent
compliance-icons
Product Identifier
  • MZ/NH3/CN
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.