missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Tissue Non-Specific Antibody (2F4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-22193
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Tissue Non-Specific Monoclonal specifically detects Alkaline Phosphatase, Tissue Non-Specific in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Alkaline Phosphatase, Tissue Non-Specific | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| ALPL | |
| Purified recombinant fragment of human Alkaline Phosphatase, Tissue Non-Specific expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 2F4 | |
| Unconjugated | |
| Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 | |
| Mouse | |
| 57.3 kDa | |
| 0.1 mL | |
| Embryonic Stem Cell Markers, Protein Phosphatase, Stem Cell Markers | |
| 249 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction