missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Alkaline Phosphatase (PAP)

Product Code. 16052545
Change view
Click to view available options
Quantity:
1000 units
Unit Size:
1000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16052545 1000 units 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16052545 Supplier Abnova™ Supplier No. D0009.1000Units
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

E.coli harboring the plasmid encoding the gene of alkaline phosphatase from Shewanella sp. SIB1 (PAP).

Specifications

Concentration 5 U/μL
For Use With (Application) Cloning
Quantity 1000 units
Includes 10 x Alkaline Phosphatase Reaction Buffer, Dilution Buffer (1 x Reaction Buffer). Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.