missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Liver/Bone/Kidney Antibody (BGN/03/662), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66385H
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Liver/Bone/Kidney Monoclonal specifically detects Alkaline Phosphatase, Liver/Bone/Kidney in Human, Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Alkaline Phosphatase, Liver/Bone/Kidney | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ALPL | |
| Alkaline phosphatase from human bone. | |
| 0.1 mL | |
| Primary | |
| NB100-66385 recognizes bone specific alkaline phosphatase. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| BGN/03/662 | |
| Western Blot, ELISA | |
| ALPL, BALP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 249 | |
| Human, Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction