missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-7328-100ug
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Intestinal Polyclonal antibody specifically detects Alkaline Phosphatase, Intestinal in Bovine samples. It is validated for Western Blot, ELISA
Specifications
| Alkaline Phosphatase, Intestinal | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:4000-1:20000 | |
| P19111 | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Reconstitute with 100 μg deionized water (or equivalent) | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
| Alkaline Phosphatase, Intestinal, Intestinal [Calf Intestine] (Uniprot: P19111) | |
| 100 μg | |
| Stem Cell Markers | |
| 248 | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction