missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline phosphatase from E. coli, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 kU
Unit Size:
5000 units
Description
- It is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection.
- It may be used for protein labeling when high sensitivity is required.
- Alkaline phosphatase is a hydrolase enzyme responsible for removing phosphate groups from many types of molecules, including nucleotides, proteins, and alkaloids.
- It is most effective in an alkaline environment. It is sometimes used synonymously as basic phosphatase.
Specifications
Specifications
| pH | 10.4 |
| For Use With (Application) | Cell Biology and Analysis, Molecular biology |
| Content And Storage | Store at 4°C, do not freeze. |
| Source | E.coli |
| Quantity | 5 kU |
| Product Type | Alkaline Phosphatase |
| Synonym | Orthophosphoric Monoester Phosphohydrolase |
| Form | Suspension |
Safety and Handling
Product Identifier
- ALKALINE PHOSPHATASE
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Ammonium Sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction