missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Bacterial Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-500-100ug
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Bacterial Polyclonal antibody specifically detects Alkaline Phosphatase, Bacterial in E. coli samples. It is validated for Western Blot, ELISA
Specifications
| Alkaline Phosphatase, Bacterial | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| BAP, ECK0378, JW0374, Pho A, PhoA | |
| Alkaline Phosphatase, Bacterial, Bacterial [E.coli] (Uniprot: P00634) | |
| 100 μg | |
| Fusion Proteins, Stem Cell Markers | |
| 945041 | |
| E. coli | |
| Purified |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 1:500-1:2000, ELISA 1:4000-1:20000 | |
| P00634 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Primary | |
| Reconstitute with 100 μg deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction