missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (HD 11F7), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Alkaline Phosphatase/ALPP Monoclonal antibody specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA, Immunohistochemistry, Radioimmune Assays
Specifications
Specifications
| Host Species | Mouse |
| Purification Method | Protein A purified |
| Research Discipline | Cancer, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?