missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (HD 11F7) - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay.
Specifications
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
| Applications | ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Immunohistochemistry (Frozen), Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | HD 11F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry : 1:10 - 1:500, Radioimmunoassay : 1:100 - 1:2000 |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme |
| Gene Symbols | ALPP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?