missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Phosphatase/ALPP Antibody (HD 11F7), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18367230 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367230 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367230 Supplier Novus Biologicals Supplier No. NBP276654AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alkaline Phosphatase/ALPP
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HD 11F7
Conjugate Alexa Fluor 488
Dilution ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme
Gene Symbols ALPP
Host Species Mouse
Immunogen Fresh Hep-2 cells followed by booster injections of fresh Hep-2 cells that had their surface expression of Alkaline Phosphatase/ALPP boosted in culture.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 250
Test Specificity This antibody recognizes both Regan and Nagao isoenzymes of placental alkaline phosphatase (PLAP).
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.