missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (H17E2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-750H
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Alkaline Phosphatase/ALPP | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ALPP | |
| Placental membrane (Human). | |
| 0.1 mL | |
| Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers | |
| 250 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| H17E2 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognises human placental alkaline phosphate. The has been widely used in immunolocalisation studies of germ cell tumours and may also be used in ELISA based assays for the measurement of soluble PLAP. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction