missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (8B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB110-3638SS
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunodiffusion.
Specifications
| Alkaline Phosphatase/ALPP | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Flow Cytometry reported in multiple pieces of scientific literature, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunodiffusion | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| Mouse | |
| 70 kDa | |
| 0.025 mL | |
| Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 250 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8B6 | |
| Unconjugated | |
| P05187 | |
| ALPP | |
| Hep-2 cells with boosted surface expression of Alkaline Phosphatase, Placental. [UniProt# P05187]. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Alkaline Phosphatase, Placental - both Regan and Nagao isoenzymes. No cross reactivity with other isoenzymes of Alkaline Phosphatase. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction