missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPL Rabbit anti-Human, Clone: 71, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89224-50UL
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Western Blot.
Specifications
| Alkaline Phosphatase/ALPL | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 71 | |
| Western Blot 1:500-1:2000 | |
| Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 | |
| A synthetic peptide corresponding to the center region of the Human Alkaline Phosphatase/ALPL | |
| 50 μg | |
| Stem Cell Markers | |
| 249 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction