missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Phosphatase/ALPL Antibody (928929) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30731818 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731818 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731818 Supplier Novus Biologicals Supplier No. FAB29092I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Alkaline Phosphatase/ALPL Monoclonal antibody specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alkaline Phosphatase/ALPL
Applications Immunohistochemistry, Western Blot, Neutralization
Classification Monoclonal
Clone 928929
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Alkaline Phosphatase/ALPL, Leu18-Ser502, Accession # P05186
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 249
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.