missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Phosphatase/ALPL Antibody (071), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18376340 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376340 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376340 Supplier Novus Biologicals Supplier No. NBP289224AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alkaline Phosphatase/ALPL
Applications Western Blot
Classification Monoclonal
Clone 71
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Alkaline Phosphatase/ALPL.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 249
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.