missing translation for 'onlineSavingsMsg'
Learn More

AGMA Alkaline Detergent

Product Code. 30066728
Change view
Click to view available options
Container Type:
Capped Bottle
Trigger Spray
Quantity:
4 x 5 L
15 x 1 L
Unit Size:
Pack of 15
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Quantity unitSize
30066728 Trigger Spray 15 x 1 L Pack of 15
30066762 Capped Bottle 4 x 5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30066728 Supplier AGMA Supplier No. 0089
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Protect your cleanrooms with a highly-effective, deep cleaning Alkaline detergent

Alkaline Detergent effectively removes stubborn contaminants prior to disinfection, including visible soiling or build-up of residue from previous disinfectant applications.

With a pH 12, it is highly effective at breaking down proteins, which makes it perfect for the first step in cleaning up spills or for preliminary cleans of new manufacturing cleanrooms.

  • pH 12 ideal for deep cleans and high protein load cleaning
  • Particularly effective against proteinaceous and organic residues
  • Removes build-up that neutral or acidic cleaners may not
  • Low foaming
  • Does not leave significant residues
  • Made with WFI water
  • Manufactured in Class C/D cleanrooms
  • Quality Management Certified to BS EN ISO 9001:2015
  • Manufactured in accordance with the cGMP Guidelines
  • Fully traceable with supporting analysis
  • Available in 1 L Trigger Sprays and 5 L capped bottled
  • Recommended for use within Class C or D cleanrooms (in line with your cGMP risk assessments, SOPs and validations)

Specifications

Product Type Alkaline Detergent in WFI
Color-coded No
Form Liquid Spray
pH 11.5 to 12.5
Scent Unscented
Sterility Non-sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Container Type Trigger Spray
Disposable Limited-use
Filtered Filtered
Flammable No
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Quantity 15 x 1 L
Show More Show Less
compliance-icons
Product Identifier
  • Alkaline Detergent
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-BUTOXYETHOXY)ETHANOL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.