missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd Alkaline Cyanide Reagent

Product Code. 15989236
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15989236 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15989236 Supplier Hach Pacific Pty Ltd Supplier No. 2122349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of low level manganese by the PAN method.

For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.

    Specifications

  • Method Name: PAN
  • Platform: Solution
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 500 mL
Type Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.