missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Ceramidase 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Alkaline Ceramidase 2 Polyclonal specifically detects Alkaline Ceramidase 2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Alkaline Ceramidase 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Acylsphingosine deacylase 3-like, Alkaline CDase 2, alkaline ceramidase 2, AlkCDase 2, ALKCDase2, ASAH3L, ceramide hydrolase, EC 3.5.1.23, FLJ41587, haCER2, N-acylsphingosine amidohydrolase 3-like |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human Alkaline Ceramidase 2 (NP_001010887). Peptide sequence DELAVLWVLMCALAMWFPRRYLPKIFRNDRGRFKVVVSVLSAVTTCLAFV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?