Learn More
Eurofins DiscoverX Alk Protein, Active
Description
ALK or anaplastic lymphoma kinase (Ki-1) is a membrane-spanning tyrosine kinase receptor that is most closely related to leukocyte tyrosine kinase. No function has been ascribed for ALK; however, its restricted expression in neural tissue suggests that this receptor might play an important role in the normal development and function of the nervous system. Large-cell anaplastic lymphoma is a subtype of non-Hodgkin's lymphoma. Over half of these lymphomas have a chromosomal translocation t(2;5), that leads to the expression of a hybrid protein comprised of the nucleolar phosphoprotein nucleophosmin (NPM) and ALK.
GenBank U62540
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 63.8 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥50% |
| Protein | Alk, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.