missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK Mouse, Clone: ALK/1503, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | ALK |
|---|---|
| Clone | ALK/1503 |
| Concentration | 200μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Specifications
| ALK | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 238 | |
| Recombinant human ALK protein fragment (aa1360-1460) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 80kDa (hybrid) ;200kDa (wild type) |
| ALK/1503 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_004295 | |
| ALK | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic and nuclear | |
| Anaplastic Large Cell Lymphoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title