missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALK Monoclonal Antibody (K40048_8D10), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | ALK |
|---|---|
| Clone | K40048_8D10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30194594
|
Invitrogen™
MA556165 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30201879
|
Invitrogen™
MA555785 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: ISVRVPRGPA VEGGHVNMAF SQSNPPSELH KVHGSRNKPT SLWNPTYGSW FTEKPTKKNN PIAKKEPHDR
ALK Monoclonal antibody specifically detects ALK in Human samples. It is validated for Western BlotSpecifications
| ALK | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Alk; ALK tyrosine kinase receptor; ALK/NPM1 fusion gene; anaplastic lymphoma kinase; Anaplastic lymphoma kinase Ki1; Anaplastic Lymphoma Kinase p80; anaplastic lymphoma receptor tyrosine kinase; CD246; CD246 antigen; EC 2.7.10.1; kinase ALK; mutant anaplastic lymphoma kinase; NBLST3; npm-alk; p80; Tcrz; TFG/ALK | |
| Recombinant human ALK (immunogen range is 1461-1530). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ALK |
| K40048_8D10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UM73 | |
| 238 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title