missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Mouse anti-Human, Clone: MM0015-8G33, Novus Biologicals™
Mouse Monoclonal Antibody
£423.00
Specifications
| Antigen | ALK/CD246 |
|---|---|
| Clone | MM0015-8G33 |
| Dilution | Western Blot 1:200-1:800, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18573132
|
Novus Biologicals
NB110-85462 |
0.1 mg | |||||||
Description
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ALK/CD246 | |
| Western Blot 1:200-1:800, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Lyophilized | |
| RUO | |
| Human | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
| human TGF RI (ALK1) extra-cellular domain | |
| Primary | |
| Reconstitute to desired concentration. |
| MM0015-8G33 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase, Signal Transduction | |
| A 0.2 um filtered solution in PBS | |
| 238 | |
| IgG2 | |
| Protein G purified | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title