missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Antibody (ALK/1504), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08446IR
This item is not returnable.
View return policy
Description
ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ALK/CD246 | |
| Monoclonal | |
| DyLight 755 | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
| Recombinant human ALK/CD246 protein fragment (aa200-335) (Uniprot: Q9UM73 ) | |
| 100 μg | |
| Protein Kinase, Signal Transduction | |
| 238 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| ALK/1504 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction