missing translation for 'onlineSavingsMsg'
Learn More

ALK/CD246 Antibody (ALK/1031), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18363762 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363762 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363762 Supplier Novus Biologicals Supplier No. NBP308771JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALK/CD246
Applications Western Blot
Classification Monoclonal
Clone ALK/1031
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Host Species Mouse
Immunogen Recombinant human ALK/CD246 protein fragment covering ∼aa400-500.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 238
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.