missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Antibody (ALK/1031), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08771CL1
This item is not returnable.
View return policy
Description
ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Western Blot
Specifications
| ALK/CD246 | |
| Monoclonal | |
| CoraFluor 1 | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
| Recombinant human ALK/CD246 protein fragment covering ∽aa400-500. (Uniprot: Q9UM73) | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 238 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Western Blot | |
| ALK/1031 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction