missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Yellow R sodium salt, Thermo Scientific Chemicals
£43.10 - £56.50
Chemical Identifiers
CAS | 1718-34-9 |
---|---|
Molecular Formula | C13H8N3NaO5 |
Molecular Weight (g/mol) | 309.213 |
MDL Number | MFCD00067121 |
InChI Key | HKSANXZSKDUDBD-UHFFFAOYSA-M |
Synonym | alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt, |
PubChem CID | 44135758 |
IUPAC Name | sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate |
SMILES | C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358977
|
Thermo Scientific Alfa Aesar
038707.09 |
10 g |
£43.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368977
|
Thermo Scientific Alfa Aesar
038707.14 |
25 g |
£56.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1718-34-9 | |
309.213 | |
HKSANXZSKDUDBD-UHFFFAOYSA-M | |
44135758 | |
C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+] |
C13H8N3NaO5 | |
MFCD00067121 | |
alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt, | |
sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate |
Specifications
1718-34-9 | |
Powder | |
MFCD00067121 | |
alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt, | |
C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+] | |
309.213 | |
309.22 | |
Dark yellow to brown powder |
Brown to Yellow | |
C13H8N3NaO5 | |
962365 | |
HKSANXZSKDUDBD-UHFFFAOYSA-M | |
sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate | |
44135758 | |
Odorless | |
Alizarin Yellow R sodium salt |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 217-002-4
missing translation for 'rtecsNumber' : DH2528600
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only