missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Yellow R sodium salt

Product Code. 11368977
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368977 25 g 25g
11358977 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368977 Supplier Thermo Scientific Alfa Aesar Supplier No. 038707.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1718-34-9
Molecular Formula C13H8N3NaO5
Molecular Weight (g/mol) 309.213
MDL Number MFCD00067121
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
Synonym alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt,
PubChem CID 44135758
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

Color Brown to Yellow
Physical Form Powder
Quantity 25 g
Beilstein 962365
Formula Weight 309.22
Odor Odorless
Appearance Dark yellow to brown powder
Chemical Name or Material Alizarin Yellow R sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.