missing translation for 'onlineSavingsMsg'
Learn More

Alizarin, TRC

Product Code. 30010182
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010182 500 mg 500mg
30012346 10 g 10g
30009389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010182 Supplier Toronto Research Chemicals Supplier No. TRCA536600500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-48-0
Molecular Formula C14 H8 O4
Molecular Weight (g/mol) 240.21
Synonym 9,10-Anthracenedione, 1,2-dihydroxy-, Alizarin B (6CI), Anthraquinone, 1,2-dihydroxy- (8CI), 1,2-Anthraquinonediol, 1,2-Dihydroxy-9,10-anthracenedione, 1,2-Dihydroxy-9,10-anthraquinone, 1,2-Dihydroxyanthraquinone, Acid Metachrome Red B, Acid Mordant Red B, Alizarin, Alizarin Red, Alizarina, Alizarine, Alizarine 3B, Alizarine B, Alizarine Indicator, Alizarine L Paste, Alizarine Lake Red 2P, Alizarine Lake Red 3P, Alizarine Lake Red IPX, Alizarine NAC, Alizarine Paste 20 percent Bluish, Alizarine Red, Alizarine Red B, Alizarine Red B2, Alizarine Red IP, Alizarine Red IPP, Alizarine Red L, C Ext. Red 62, C.I. 58000, C.I. Mordant Red 11, Certiqual Alizarine, D And C Orange Number 15, Deep Crimson Madder 10821, Eljon Madder, Mitsui Alizarine B, Mordant Red 11, NSC 7212, Qiansu, Turkey red
IUPAC Name 1,2-dihydroxyanthracene-9,10-dione
SMILES Oc1ccc2C(=O)c3ccccc3C(=O)c2c1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H8O4/c15-10-6-5-9-11(14(10)18)13(17)8-4-2-1-3-7(8)12(9)16/h1-6,15,18H
Formula Weight 240.0423
Chemical Name or Material Alizarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.