missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S sodium salt

Product Code. 11319707
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11319707

Brand: Thermo Scientific Alfa Aesar 042040.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transition interval: pH 4.6 (yellow) to pH 6.0 (red) | CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

Alizarin Red S sodium salt is used as a chromogenic agent for the colorimetric determination of dothiepin hydrochloride in pharmaceutical formulations. It is also used as an ion-pair reagent for the spectrophotometric assay of fexofenadine in pharmaceuticals. It acts as a colorometric pH indicator in the range of 3.8-5.4. Further, it is used in a biochemical assay to determine the presence of calcific deposition by cells of an osteogenic lineage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
Synonym alizarin red s, sodium alizarinesulfonate, mordant red 3, sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, alizarine s, acid red alizarine, sodium alizarin-3-sulfonate, alizarine red s sodium salt, alizarinsulfonate, alizarin s
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Physical Form Powder
Quantity 5 g
Beilstein 4117090
Solubility Information Soluble in water and alcohol.
Formula Weight 342.27
Appearance Various color powder
Chemical Name or Material Alizarin Red S sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.