missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin Red S sodium salt, 1% w/v aq. sol.

Product Code. 11366149
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366149 500 mL 500mL
11345547 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366149 Supplier Thermo Scientific Chemicals Supplier No. 042746.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transition interval: pH 4.6 (yellow) to pH 6.0 (red) | CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Formula Weight 342.27
Odor Odorless
Color Amber
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Alizarin Red S sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.