missing translation for 'onlineSavingsMsg'
Learn More

Alizarin complexone, MP Biomedicals™

Product Code. 11451975 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451975 Supplier MP Biomedicals Supplier No. 0520342901
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alizarin complexone is suitable for the colorimetric detection of fluorine. Alizarin complexone is also known that it has antiretroviral activity for the medication purpose of the treatment of infection by retroviruses

Specifications

CAS 3952-78-1
Molecular Formula C19H15NO8
MDL Number MFCD00001202
Synonym Alizarin-3-methylamine-N, N-diacetic acid
Color Brown
Quantity 1 g
InChI Key PWIGYBONXWGOQE-UHFFFAOYSA-N
Physical Form Fine Powder
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O
IUPAC Name 2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
Molecular Weight (g/mol) 385.328
PubChem CID 65132
ChEBI CHEBI:53088
Formula Weight 385.34
Grade Not Available
Chemical Name or Material Alizarin complexone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.