Learn More
Alix Monoclonal Antibody (3A9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA183977
Code : LE
Additional Details : Weight : 0.10000kg
Description
This product does not require protein digestion pre-treatment of paraffin sections. A ∽100kD band is detected by Western blot. Mouse anti ALIX antibody, clone 3A9 recognizes the apoptosis-linked gene-2 interacting protein X (ALIX), also known as Programmed cell death 6-interacting protein or Hp95.
Apoptotic linked-gene-product 2 (ALG-2) interacting protein X (ALIX) is a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral budding and actin cytoskeleton assembly.Specifications
Alix | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q8WUM4, Q9QZA2, Q9W6C5, Q9WU78 | |
Pdcd6ip, pdcd6ip.L | |
Alix fusion protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat, X. laevis | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
3A9 | |
Unconjugated | |
Pdcd6ip | |
AI480591; AIP1; ALG-2 interacting protein 1; ALG-2 interacting protein X; ALG-2-interacting protein 1; Alg2-interacting protein 1; ALG-2-interacting protein X; Alg2-interacting protein X; Alix; apoptosis-linked gene 2-interacting protein X; AW544830; C76364; dopamine receptor interacting protein 4; DRIP4; E2F1-inducible protein; Eig2; Hp95; KIAA1375; MGC17003; mKIAA1375; PDCD6-interacting protein; Pdcd6ip; pdcd6ip.L; programmed cell death 6 interacting protein; programmed cell death 6 interacting protein L homeolog; programmed cell death 6-interacting protein; putative signal tranduction protein Xp95; RGD1561176; Signal transduction protein Xp95; XELAEV_18031317mg; xp95 | |
Mouse | |
Protein A | |
RUO | |
10015, 18571, 398095, 501083 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |