missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alix Antibody (3A9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
£203.00 - £498.00
Specifications
| Antigen | Alix |
|---|---|
| Clone | 3A9 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18789923
|
Novus Biologicals
NB100-65678SS |
0.025 mg |
£203.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761134
|
Novus Biologicals
NB100-65678 |
0.1 mg |
£498.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alix Monoclonal specifically detects Alix in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Alix | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Xenopus species | |
| AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein | |
| PDCD6IP | |
| IgG1 | |
| Protein G purified | |
| NB100-65678 recognizes apoptotic-linked-gene-product 2 (ALG-2) interacting protein X (ALIX), a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral budding and actin cytoskeleton assembly. |
| 3A9 | |
| Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q8WUM4 | |
| 10015 | |
| Alix fusion protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title