missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.8900 g/mL |
| Flash Point | 77°C |
| Quantity | 2.5 L |
| Specific Gravity | 0.89 |
| Solubility Information | Solubility aq. soln.: insoluble. |
| Viscosity | 1500 mPa.s (30°C) |
| Chemical Name or Material | Aliquat* 336 TG |
Safety and Handling
Product Identifier
- Aliquat 336TG
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Quaternary ammoniumcompounds, tri-C8-10-alkylmethyl, chlorides, 1-Octanol, 1-Decanol
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?