missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid, TRC

Product Code. 17919104
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17919104 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919104 Supplier Toronto Research Chemicals Supplier No. TRCA5330001G
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9005-32-7
Molecular Formula [C6H8O6]n
Quantity 1 g
Synonym A 2830-9, A 7003, Acid Algin G 2, Alginate 8, Alginate LV, Cecalgum S 500, Duck Acid A, Duckacid X 2787, E 400, Grindsted PH 060, Inagel GS 30, Kelacid, Kimica Acid, Kimica Acid G, Kimica Acid R 1, Laminal, Lamitex LV, Landalgine, MediAlg MG 17, Medialg MG 5, Medisorb A, Norgine, Protacid F 120, Protanal Rf 5650, Protanal Sf 120RB, Satialgin S 1100, Satialgine, Satialgine GCF 645, Satialgine H 8, Snow Acid Algin G, Verdyol Super
InChI Formula InChI=1S/C14H22O13/c1-23-7-3(15)6(18)14(27-9(7)11(19)20)25-8-4(16)5(17)13(24-2)26-10(8)12(21)22/h3-10,13-18H,1-2H3,(H,19,20)(H,21,22)/t3-,4-,5+,6+,7+,8+,9-,10+,13-,14-/m1/s1
IUPAC Name (2S,3S,4R,5S,6R)-3-(((2R,3S,4R,5S,6R)-6-carboxy-3,4-dihydroxy-5-methoxytetrahydro-2H-pyran-2-yl)oxy)-4,5-dihydroxy-6-methoxytetrahydro-2H-pyran-2-carboxylic acid
Chemical Name or Material Alginic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.