missing translation for 'onlineSavingsMsg'
Learn More

Alginic acid

Product Code. p-7023735
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11458207

Brand: Thermo Scientific Alfa Aesar A17582.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B

A straight chain, hydrophilic, colloidal polyuronic acidAlginic acid is used as a precursor for the preparation of sodium alginate, potassium alginate and calcium alginate, which finds application in various industries such as food, textile printing and pharmaceuticals. It acts as an additive in dehydrated products such as slimming aids. It is also used in the production of paper and textiles. It is actively involved in waterproofing, fireproofing fabrics. It is utilized as a thickening agent in drinks, ice cream and cosmetics. It plays an important role as a raw material for the synthesis of propylene glycol alginate and polysaccharide sulfate. It is also useful to treat and purify waste water having strong absorbility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-32-7
Molecular Formula (C6H8O7)A(C6H8O7)B
MDL Number MFCD00081309
Synonym 2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid

Specifications

Melting Point >300°C
Merck Index 14,242
Solubility Information Insoluble in water.
Quantity 100 g
Sensitivity Hygroscopic
Chemical Name or Material Alginic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.