missing translation for 'onlineSavingsMsg'
Learn More

Alginic acid sodium salt, very low viscosity

Product Code. 11428497
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11428497

Brand: Thermo Scientific Alfa Aesar A18565.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

It is a hematologic agent, drug that acts on blood and blood-forming organs and those that affect the hemostatic system. sodium alginate has been used in warp sizing, packing and printing. In printing, it is an ideal kind of material with wide applications in the cotton, wool, silk and synthetic fiber. Sodium alginate is widely used in food industry such as it is the main material of seaweed, synthetic grape and synthetic cherry, it is taken as a stabilizer of ice cream and ice cream bar because of its dense organization and slow solution speed. It is also the main material of the dainty cold powder and jelly, As a stabilizer of cake (such as cookies, bread, fine dried noodle, chocolates) and a polish of bread, sodium alginate can keep the cookies sweet-smelling and avoid falling to pieces, and make the noodle smooth and reduce breaking ratio of noodle and fragmentation of bread. Sodium alginate is also a good thickener of jam, chili sauce, jelly, tomato ketchup, fishpaste, pudding and salad flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin; Polymannuronic acid sodium salt

Specifications

Merck Index 14,241
Solubility Information Soluble in water.
Quantity 500 g
Sensitivity Hygroscopic
Chemical Name or Material Alginic acid sodium salt, very low viscosity

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.