missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid Sodium Salt, Technical Grade, TRC

Product Code. 17938134
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17938134 100 g 100g
17949104 5 g 5g
17917184 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938134 Supplier Toronto Research Chemicals Supplier No. TRCA533005100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6 H7 O6 Na)n
Molecular Weight (g/mol) 216.12
Synonym Algin, Sodium alginate
IUPAC Name sodium;(2S,3S,4S,5S,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES [C@@H]1([C@@H]([C@H](O[C@H]([C@H]1O)O)C(=O)[O-])O)O.[Na+]

Specifications

Quantity 100 g
InChI Formula InChi=1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1-,2-,3-,4?,6+;/m0./s1
Chemical Name or Material Sodium alginate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.