missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid, Sodium Salt, Medium Viscosity, MP Biomedicals

Product Code. 11464751 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464751 100 g 100g
11474751 250 g 250g
11484751 500 g 500g
11494751 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11464751 Supplier MP Biomedicals Supplier No. 0215472480
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Source: kelp

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
Synonym Sodium alginate

Specifications

Color White to Cream
Physical Form Powder
Quantity 100 g
Assay Percent Range >98%
Linear Formula (C6H7O6Na)n
Chemical Name or Material Alginic Acid, Sodium Salt, Medium Viscosity, Viscosity (1% aq. solution): 600-900 cps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.