missing translation for 'onlineSavingsMsg'
Learn More

Alginic acid sodium salt, low viscosity

Product Code. p-7028126
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11410828

Brand: Thermo Scientific Alfa Aesar B25266.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

A gelling polysaccharide extracted from giant brown seaweed, it is useful in viscosity studiesAlginic acid sodium salt is widely utilized in the food industry as flavorless gum which enhances viscosity and acts as an emulsifier. It finds application as a thickener for reactive dyestuffs which is used in textile screen printing and carpet jet-printing. It is used as indigestion tablets and for the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold which is useful for tissue engineering applications. It acts as a food additive and is used in production of gel-like foods such as baking. It is also used for encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin; Polymannuronic acid sodium salt

Specifications

Merck Index 14,241
Solubility Information Soluble in water. Insoluble in alcohol,chloroform and ether.
Quantity 100 g
Sensitivity Hygroscopic
Chemical Name or Material Alginic acid sodium salt, low viscosity

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.