missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlgiMatrix™ 6-Well, 3D Culture System, Flat-Bottom Microplate

Product Code. 10360684
Click to view available options
Quantity:
1 x 6 well plate
4 x 6 well plates
No. of Wells:
6
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10360684

Brand: Gibco™ A1098201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Color: Clear; Sterility: Sterile; Lid: With Lid

AlgiMatrix™ 3D Culture System 6 well plate is a user-friendly, animal –free bioscaffold available for the development of high-fidelity cell culture models that are more predictive of disease states and drug responses. AlgiMatrix™ sponges are formulated using pharmaceutical-grade raw material from brown seaweed. Its pure, chemically defined, and nontoxic macro porous structure improved lot-to-lot consistency, superior cell loading and excellent nutrient delivery, without damage to cells. The AlgiMatrix™ sponge is biodegradable and is stable at room temperature.
 
The AlgiMatrix™ 3D Culture System is suitable for a broad range of procedures; including Multicellular tumor spheroid assays (MCTS), hepatocyte and cardiomyocyte organogenesis studies, co-culture studies, and high-throughput drug screening assays.
TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Content And Storage Contains 1 x 6 well AlgiMatrix™ plate and 50 ml firming buffer. Store at room temperature, 15-30° C
Product Line Algimatrix
Color Clear
Product Type Microplate
No. of Wells 6
Sterility Sterile
Surface Treatment AlgiMatrix 3D Culture System
Well Shape Flat Bottom (F-well)
Includes 1 x 6 well AlgiMatrix Plate and 50 mL Firming Buffer
Purity or Quality Grade Cell Culture Grade
Quantity 1 x 6 well plate
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.