missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlgiMatrix™ 24-Well, 3D Culture System, Flat-Bottom Microplate

Product Code. 10761574
Click to view available options
Quantity:
1 x 24 well plate
4 x 24 well plates
No. of Wells:
24
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10761574

Brand: Gibco™ 12684049

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Color: Clear; Sterility: Sterile; Lid: With Lid

The Algimatrix™ 3D Culture System provides a three-dimensional matrix that has the potential to create cell culture conditions that more closely reflect normal cell morphology and behavior. It has relevance for both academic and industrial researchers who want to culture hepatocytes and other spheriod forming cells. Algimatrix™ alginate sponge is provided in a 24 well format that fits well into the workflow of both lower and higher-throughput users. The larger well size enables growth of more cells for downstream applications such as transfection.
TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Content And Storage Higher-fidelity, animal-free scaffold for 3D cell culture that mimics in vivo conditions. More consistent results than animal-derived matrices. Ready to use, stored at room temperature and stable up to 18 months. Protocols available.
Packaging 4 Pack
Product Line Algimatrix
Color Clear
Product Type Microplate
No. of Wells 24
Sterility Sterile
Surface Treatment AlgiMatrix 3D Culture System
Well Shape Flat Bottom (F-well)
Purity or Quality Grade Cell Culture Grade
Quantity 4 x 24 well plates
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.