missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALG9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109381
This item is not returnable.
View return policy
Description
Immunogen sequence: LFRGYHGPLD LYPEFYRIAT DPTIHTVPEG RPVNVCVGKE WYRFPSSFLL PDNWQLQFIP SEFRGQLPKP FAEGPLATRI VPTDMNDQNL EEPSRYIDIS KCHYLVDLDT MRETPREPKY SSNKEEWISL AYRPFLDASR SSKLLRAFYV PFLSDQYTVY VNYTILKPRK AKQIRKKSGG.
This gene encodes an alpha-1,2-mannosyltransferase enzyme that functions in lipid-linked oligosaccharide assembly. Mutations in this gene result in congenital disorder of glycosylation type Il. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ALG9 | |
| Polyclonal | |
| Unconjugated | |
| ALG9 | |
| ALG9; ALG9, alpha-1,2-mannosyltransferase; alpha-1,2-mannosyltransferase ALG9; asparagine-linked glycosylation 9 alpha-12-mannosyltransferase-like protein; asparagine-linked glycosylation 9 homolog (S. cerevisiae, alpha- 1,2-mannosyltransferase); asparagine-linked glycosylation 9 homolog (yeast, alpha- 1,2-mannosyltransferase); asparagine-linked glycosylation 9, alpha-1,2-mannosyltransferase homolog; asparagine-linked glycosylation protein 9 homolog; CDG1L; DIBD1; disrupted in bipolar affective disorder 1; disrupted in bipolar disorder protein 1; dolichyl-P-Man:Man(6)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; dolichyl-P-Man:Man(8)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; dol-P-Man dependent alpha-1,2-mannosyltransferase; dol-P-Man:Man(6)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; Dol-P-Man:Man(8)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; GIKANIS; LOH11CR1J; loss of heterozygosity, 11, chromosomal region 1 gene J product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79796 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.78 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9H6U8 | |
| ALG9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 439-618 of human ALG9 (NP_079016.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction