Learn More
ALG9 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29218-1-AP-150UL
This item is not returnable.
View return policy
Description
The ALG9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ALG9. This antibody recognizes human antigen. The ALG9 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ALG9 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ALG9, DIBD1, DKFZp586M2420, FLJ21845, LOH11CR1J | |
| 150 μL | |
| 79796 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |