missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88767
This item is not returnable.
View return policy
Description
ALG5 Polyclonal specifically detects ALG5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ALG5 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Q9Y673 | |
| ALG5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RHEEEKFFLNAKGQKETLPSIWDSPTKQLSVVVPSYNEEKRLPVMMDEALSYLEKRQKRDPAFTYEVIVVDDGSKDQTSKVAFKYCQKYGSDKVRVITLVKNRGKGGAIRMGIFSSRGEKILMADADGATKFPDVEKLEKGLNDL | |
| Affinity Purified | |
| RUO | |
| 29880 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Alg5, S. cerevisiae, homolog of, asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphatebeta-glucosyltransferase), asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphatebeta-glucosyltransferase), asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferasehomolog (S. cerevisiae), Asparagine-linked glycosylation protein 5 homolog, bA421P11.2, dolichyl phosphate glucosyltransferase, dolichyl-phosphate beta-glucosyltransferase, DolP-glucosyltransferase, EC 2.4.1, EC 2.4.1.117, RP11-421P11.2 | |
| Rabbit | |
| 37 kDa | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction