missing translation for 'onlineSavingsMsg'
Learn More

ALG5 Antibody, Novus Biologicals™

Product Code. 18361030 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361030 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361030 Supplier Novus Biologicals Supplier No. NBP286978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ALG5 Polyclonal specifically detects ALG5 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias Alg5, S. cerevisiae, homolog of, asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphatebeta-glucosyltransferase), asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphatebeta-glucosyltransferase), asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferasehomolog (S. cerevisiae), Asparagine-linked glycosylation protein 5 homolog, bA421P11.2, dolichyl phosphate glucosyltransferase, dolichyl-phosphate beta-glucosyltransferase, DolP-glucosyltransferase, EC 2.4.1, EC 2.4.1.117, RP11-421P11.2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of ALG5
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29880
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.