missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALG1 Polyclonal Antibody

Product Code. 12848062
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848062 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848062 Supplier Invitrogen™ Supplier No. PA1031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ALG1 Polyclonal Antibody for Western Blot

The enzyme encoded by this gene catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. This gene is mutated in congenital disorder of glycosylation type Ik.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene alg-1
Gene Accession No. 0
Gene Alias Alg1; alg-1; ALG1 chitobiosyldiphosphodolichol beta-mannosyltransferase; ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase; Argonaute; Argonaute-like; asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (S. cerevisiae, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; beta-1,4-mannosyltransferase; CDG1K; CELE_F48F7.1; chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; HMAT1; hMat-1; HMT1; HMT-1; Mannosyltransferase-1; Mat-1; MT-1; Protein argonaute; protein-coding; PSEC0061; UNQ861/PRO1870
Gene Symbols alg-1
Host Species Rabbit
Immunogen Peptide sequence DLEEIPNSPPTQPQTFSDVPQR.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 181504
Target Species C. elegans
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
compliance-icons
Product Identifier
  • ALG1 Polyclonal Antibody
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.