missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | ALG1 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18438751
|
Novus Biologicals
NBP2-32539-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18126022
|
Novus Biologicals
NBP2-32539 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALG1 Polyclonal specifically detects ALG1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ALG1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56052 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NWHIRAVTVYDKPASFFKETPLDLQHRLFMKLGSMHSPFRARSEPEDPVTERSAFTERDAGSGLVTRLRERP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase), asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 1 homolog, beta-1,4 mannosyltransferase, beta-1,4-mannosyltransferase, chitobiosyldiphosphodolichol beta-mannosyltransferase, GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase, GDP-mannose-dolichol diphosphochitobiose mannosyltransferase, hMat-1, HMAT1EC 2.4.1.142, HMT-1, HMT1CDG1K, Mannosyltransferase-1, MT-1 | |
| ALG1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title