missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin Hydrochloride, TRC

Product Code. 30006848
Change view
Click to view available options
Quantity:
5 mg
100 mg
500 mg
Unit Size:
100mg
500mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006848 100 mg 100mg
17914825 500 mg 500mg
17970415 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006848 Supplier Toronto Research Chemicals Supplier No. TRCA532000100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81403-68-1
Molecular Formula C19 H27 N5 O4 . Cl H
Molecular Weight (g/mol) 425.91
Synonym Alfuzosin hydrochloride, (2RS)-N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]tetrahydrofuran-2-carboxamide hydrochloride, SL 77-499-10, SL 77499-10, Urion, Uroxatral, Xatral, Alfoten
IUPAC Name N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride
SMILES Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC(=O)C3CCCO3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H27N5O4.ClH/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h10-11,14H,4-9H2,1-3H3,(H,21,25)(H2,20,22,23);1H
Formula Weight 425.183
Chemical Name or Material Alfuzosin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.